Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KChIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541075 Supplier Invitrogen™ Supplier No. PA541075
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QLTDSVDDEF ELSTVCHRPE GLEQLQEQTK FTRKELQVLY RGFKNPGALS Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Voltage-gated potassium (Kv) channels are key components of the potassium currents in the heart and central nervous system. In the heart, Kv4 channels are involved in the repolarization phase of the action potential. In the brain however, these channels prevent reverse-propagation of action potentials. Found associated with Kv4 channels are a group of calcium-binding proteins termed KChIPs (Kv channel interacting proteins). KChIPs are small molecular weight proteins that bind to the cytoplasmic amino termini of Kv4 alpha-subunits and help to modulate its function. There are currently three known KChIPs; KChIP1 expressed in the brain; KChIP2 expressed in the brain, heart, and lung, and has three isoforms, a, b, and c; and KChIP3, also known as calsenilin and DREAM (downstream regulatory elements (DRE)-antagonist modulator) which is expressed in the brain and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KChIP2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNIP2
Gene Accession No. Q9JJ69, Q9NS61
Gene Alias A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit; DKFZp566L1246; KCHIP2; KChlP2; Kcnip2; Kv channel interacting protein 2; Kv channel-interacting protein; Kv channel-interacting protein 2; MGC17241; potassium channel auxiliary subunit KCHIP2; potassium channel-interacting protein 2; potassium voltage-gated channel interacting protein 2; si:ch73-173h19.2
Gene Symbols KCNIP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCNIP2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30819, 80906
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.