Learn More
Ricca VeriSpec™ KCl Conductivity Standard, Certified Reference Material
Shop All Ricca Chemical Company Products

Description
- Potassium Chloride starting materials
- Manufactured and tested in ISO 17025/17034 accredited facilities
- Certified traceable to NIST
- Wide range of concentrations
Specifications
Specifications
| Chemical Name or Material | KCl |
| Conductivity | 25,000μS/cm at 25°C |
| Technique | Calibration of Conductivity Meter |
| Color | Colorless |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| For Use With (Equipment) | Conductivity Meters and Probes |
| Solubility | Soluble |
| Solution Type | Conductivity Standard |
| Type | Certified Reference Material |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.