Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricca VeriSpec™ KCl Conductivity Standard, Certified Reference Material
SDP

Catalog No. 01670605 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Conductivity:
10 μS/cm (at 25°C)
10,000 μS/cm (at 25°C)
100 μS/cm (at 25°C)
100,000 μS/cm (at 25°C)
1000 μS/cm (at 25°C)
1413 μS/cm (at 25°C)
25,000μS/cm at 25°C
250 μS/cm (at 25°C)
447 μS/cm (at 25°C)
50 μS/cm (at 25°C)
50,000 μS/cm (50 mS/cm)
500μS/cm at 25°C
5000μS/cm at 25°C
750μS/cm at 25°C
84μS/cm at 25°C
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conductivity
01-670-605 447 μS/cm (at 25°C)
01-100-278 10 μS/cm (at 25°C)
01-185-100 25,000μS/cm at 25°C
01-670-606 50 μS/cm (at 25°C)
01-185-101 5000μS/cm at 25°C
01-185-102 750μS/cm at 25°C
01-100-281 10,000 μS/cm (at 25°C)
01-100-279 100 μS/cm (at 25°C)
01-100-280 1000 μS/cm (at 25°C)
01-100-282 100,000 μS/cm (at 25°C)
01-670-607 250 μS/cm (at 25°C)
01-670-608 1413 μS/cm (at 25°C)
01-670-604 50,000 μS/cm (50 mS/cm)
01-185-103 500μS/cm at 25°C
01-185-104 84μS/cm at 25°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 01-670-605 Supplier Ricca VeriSpec™ Supplier No. RV588747500A
Add to Cart
Add to Cart

ISO 17025/17034 KCl Conductivity Standard Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.

  • Potassium Chloride starting materials
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • Certified traceable to NIST
  • Wide range of concentrations

Specifications

Chemical Name or Material VeriSpec™ Certified Reference Material, KCl Conductivity Standard 447 μS/cm (at 25°C)
Packaging Natural Poly
CAS 7447-40-7, 7732-18-5
Traceability to NIST NIST Traceable
Product Line VeriSpec
Odor Odorless
Conductivity 447 μS/cm (at 25°C)
Physical Form Liquid
Certifications/Compliance ISO 17025, ISO 17034
Color Colorless
pH Approximately 7
Melting Point
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Industry Type Laboratory Use
Molecular Weight (g/mol) 92.56 g/mol
Quantity 500 mL
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.