Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7209585 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3543 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3543 Supplier Proteintech Group Inc Supplier No. 216591AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.

Specifications

Antigen KCNA5
Applications Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCNA5
Gene Accession No. P19024, P22460, Q61762
Gene Alias ATFB7, HCK1, HK2, HPCN1, KCNA5, KV1.5, PCN1
Gene Symbols Kcna5
Host Species Rabbit
Immunogen KCNA5 Fusion Protein Ag16397
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16493, 25470, 3741
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.