Learn More
Invitrogen™ KCNE2 Polyclonal Antibody

Description
PA1-9547 detects KCNE2 in human samples. PA1-9547 has been successfully used in Western blot procedures. By Western blot, PA1-9547 detects a 12 kDa protein representing KCNE2. PA1-9547 immunogen is a purified recombinant full length KCNE2 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | KCNE2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KCNE2 |
| Gene Accession No. | Q9Y6J6 |
| Gene Alias | 2200002I16Rik; ATFB4; AW048273; cardiac voltage-gated potassium channel accessory subunit 2; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; Kcne2; LQT5; LQT6; minimum potassium ion channel related peptide 1; minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; MIRP1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2; potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel subfamily E regulatory subunit 2; potassium voltage-gated channel, Isk-related family, member 2; potassium voltage-gated channel, Isk-related subfamily, gene 2; voltage-gated K+ channel subunit MIRP1 |
| Gene Symbols | KCNE2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.