Learn More
Invitrogen™ KCNE3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total KCNE3.
Specifications
Specifications
| Antigen | KCNE3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Kcne3 |
| Gene Accession No. | Q9JJV7, Q9WTW2, Q9Y6H6 |
| Gene Alias | 2210017H05Rik; cardiac voltage-gated potassium channel accessory subunit; HOKPP; HYPP; KCNE3; Minimum potassium ion channel-related peptide 2; minK-related peptide 2; MiRP2; potassium channel subunit beta MiRP2; potassium channel, voltage gated subfamily E regulatory beta subunit 3; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 3; potassium voltage-gated channel subfamily E member 3; potassium voltage-gated channel subfamily E regulatory subunit 3; potassium voltage-gated channel, Isk-related family, member 3; potassium voltage-gated channel, Isk-related subfamily, gene 3; potassium voltage-gated channel, Isk-related subfamily, member 3; voltage-gated K+ channel subunit MIRP2 |
| Gene Symbols | Kcne3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.