Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577042 Supplier Invitrogen™ Supplier No. PA577042
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH1 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH1 is activated at the onset of myoblast differentiation. KCNH1 is highly expressed in brain and in myoblasts. Overexpression of KCNH1 may confer a growth advantage to cancer cells and favor tumor cell proliferation. Alternative splicing of KCNH1 results in two transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNH1
Gene Accession No. O95259, Q60603, Q63472
Gene Alias EAG; EAG channel 1; EAG1; ether a go-go; ether-a-go-go potassium channel 1; ether-a-go-go, Drosophila, homolog of; h-eag; hEAG1; Kcnh1; Kv10.1; M-eag; potassium channel, voltage gated eag related subfamily H, member 1; potassium voltage-gated channel subfamily H member 1; potassium voltage-gated channel, subfamily H (eag-related), member 1; r-eag; TMBTS; voltage-gated potassium channel subunit Kv10.1; ZLS1
Gene Symbols KCNH1
Host Species Rabbit
Immunogen Recombinant full length Human KCNH1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16510, 3756, 65198
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.