Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNIP4 Mouse anti-Human, Porcine, Rat, Clone: 5B1D12, Proteintech
SDP

Catalog No. p-7212065 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6258 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6258 Supplier Proteintech Group Inc Supplier No. 601331IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.

Specifications

Antigen KCNIP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5B1D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCNIP4
Gene Accession No. Q6PIL6, Q99MG9
Gene Alias CALP, Calsenilin like protein, KCHIP4, KCNIP4
Gene Symbols KCNIP4
Host Species Mouse
Immunogen KCNIP4 Fusion Protein Ag4863
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100516499, 259243, 80333
Target Species Human, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.