Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101757 Supplier Invitrogen™ Supplier No. PA5101757
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNK1.

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kcnk1
Gene Accession No. O00180, O08581, Q9Z2T2
Gene Alias AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788)
Gene Symbols Kcnk1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNK1(Accession O00180), corresponding to amino acid residues L34-E84.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16525, 3775, 59324
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.