Learn More
Specifications
Specifications
| Gene ID (Entrez) | 56659 |
| Species | Human |
| Accession Number | NP_071337 |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
| For Use With (Application) | KCNK13 peptide is used for blocking the activity of KCNK13 antibody NBP2-41132. |
| Gene Alias | K2p13.1, potassium channel subfamily K member 13, potassium channel, subfamily K, member 13, tandem pore domain potassium channel THIK-1, THIK-1 |
| Gene Symbol | KCNK13 |
| Quantity | 0.05 mg |
| Research Category | Signal Transduction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
