Learn More
Invitrogen™ KCNK15 Polyclonal Antibody

Description
KCNK15 / KCNKF Polyclonal Antibody detects endogenous levels of KCNK15 / KCNKF protein.
Specifications
Specifications
| Antigen | KCNK15 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KCNK15 |
| Gene Accession No. | Q9H427 |
| Gene Alias | Acid-sensitive potassium channel protein TASK-5; dJ781B1.1; K2p15.1; KCNK11; KCNK14; Kcnk15; KT3.3; Potassium channel subfamily K member 15; potassium channel subfamily K member 15 (TASK-5); potassium channel, subfamily K, member 14; potassium channel, subfamily K, member 15; potassium channel, subfamily K, member 15 (TASK-5); potassium channel, two pore domain subfamily K, member 15; potassium two pore domain channel subfamily K member 15; rTASK-5; TASK5; TASK-5; TWIK-related acid-sensitive K(+) channel 5; two pore K(+) channel KT3.3; Two pore potassium channel KT3.3 |
| Gene Symbols | KCNK15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.