Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588637 Supplier Invitrogen™ Supplier No. PA588637
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGNRNSMVIH IPEEPRPSRP RYKADVPDLQ SVCSCTCYRS QDYGGRSVAP QNSFSAKLAP HYFHSISYKI EEISPSTLKN SLFPSPISSI SPGLHSFTDH QRLMKRRKSV; Positive Samples: SH-SY5Y, Mouse brain, Mouse intestine, Mouse pancreas; Cellular Location: Cell membrane, Multi-pass membrane protein.

KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Kcnk9
Gene Accession No. Q3LS21, Q9NPC2
Gene Alias acid-sensitive potassium channel protein TASK-3; K2p9.1; Kcnk9; KT3.2; MGC138268; MGC138270; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel TASK3; potassium channel, subfamily K, member 9; potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; potassium two pore domain channel subfamily K member 9; TASK3; TASK-3; TWIK-related acid-sensitive K(+) channel 3; two pore K(+) channel KT3.2; Two pore potassium channel KT3.2
Gene Symbols Kcnk9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 265-374 of human KCNK9 (NP_0012694631).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223604, 51305
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.