Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNMB4 Mouse anti-Human, Mouse, Clone: 4H7G3, Proteintech
SDP

Catalog No. p-7212367 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6250 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6250 Supplier Proteintech Group Inc Supplier No. 601221IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.

Specifications

Antigen KCNMB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4H7G3
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCNMB4
Gene Accession No. Q86W47, Q9JIN6
Gene Alias BK channel subunit beta 4, BKbeta4, Hbeta4, K(VCA)beta 4, KCNMB4, Maxi K channel subunit beta 4, Slo beta 4
Gene Symbols Kcnmb4
Host Species Mouse
Immunogen KCNMB4 Fusion Protein Ag5618
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27345, 58802
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.