Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNQ2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D4A1, Proteintech
SDP

Catalog No. 501737179 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7179 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7179 Supplier Proteintech Group Inc Supplier No. 667741IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.

Specifications

Antigen KCNQ2
Applications Western Blot
Classification Monoclonal
Clone 1D4A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCNQ2
Gene Accession No. O43526, O88943, Q9Z351
Gene Alias BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KCNQ2, KQT like 2, KV7.2, KVEBN1
Gene Symbols Kcnq2
Host Species Mouse
Immunogen KCNQ2 Fusion Protein Ag25781
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110257402, 16536, 170848, 3785
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.