Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD13 Monoclonal Antibody (OTI7D5), TrueMAB™, OriGene
SDP

Catalog No. 501682380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2380 Supplier OriGene Supplier No. TA808803
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 super family proteins are monooxygenases which catalyze multiple biochemical reactions involved in drug metabolism and synthesis of cholesterol, steroids and lipids. CYP4F3, a 60KDa protein, is a novel member of this family. It consists of a conserved heme-binding domain near the C-terminus and catalyzes the u-hydroxylation of LTB4. CYP4F3 is localized in the ER and plays a vital role in the inactivation of LTB4 and controlling inflammation. CYP4F3 exists in two functionally distinct isoforms: CYP4F3A, a neutrophil isoform, is particularly expressed in neutrophils and myeloid cells and catalyzes the microsomal u-hydroxylation of LTB4. It interacts with CD11b and myeloperoxidase on myeloid cells and regulates cellular development in bone marrow. CYP4F3B, a liver isoform, catalyzes the conversion of arachidonic acid to 20-HETE, a potent activator of PKC. Itrole hasbeen implicatedin cell proliferation, vascular tone and natriuresis.

Specifications

Antigen KCTD13
Applications Western Blot
Classification Monoclonal
Clone OTI7D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KCTD13
Gene Accession No. Q8WZ19
Gene Alias BACURD1, FKSG86, PDIP1, POLDIP1, hBACURD1
Gene Symbols KCTD13
Host Species Mouse
Immunogen Full length human recombit protein of human KCTD13 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253980
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.