Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KD Scientific Stainless-Steel Syringes

Catalog No. 1493150 Shop All BTX Harvard Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Product Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Type
14-931-50 200 mL Swagelok 1/16 in. Syringe
14-931-35 2.5 mL Swagelok 1/16 in. Syringe
14-931-36 8 mL Swagelok 1/16 in. Syringe
14-931-38 20 mL Swagelok 1/16 in. Syringe
14-931-39 20 mL Swagelok 1/8 in. Syringe
14-931-40 20 mL Swagelok 1/4 in. Syringe
14-931-37 8 mL Swagelok 1/8 in. Syringe
14-931-41 20 mL Luer-Lok Syringe
14-931-42 50 mL Swagelok 1/16 in. Syringe
14-931-43 50 mL Swagelok 1/8 in. Syringe
14-931-47 100 mL Swagelok 1/8 in. Syringe
14-931-44 50 mL Swagelok 1/4 in. Syringe
14-931-46 100 mL Swagelok 1/16 in. Syringe
14-931-49 100 mL Luer-Lok Syringe
14-931-48 100 mL Swagelok 1/4 in. Syringe
14-931-45 50 mL Luer-Lok Syringe
14-931-51 200 mL Swagelok 1/8 in. Syringe
14-931-52 200 mL Swagelok 1/4 in. Syringe
14-931-53 200 mL Luer-Lok Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 14-931-50 Supplier KD Scientific Supplier No. 780806
Add to Cart
Add to Cart

Durable, electron-beam construction withstands high force, high pressure applications

Eliminates hazards of glass syringe breakage.

  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals

Specifications

Autoclavable Autoclavable
Material Stainless Steel
Diameter (English) Inner 1-19/25 in.
Diameter (English) Outer 1-22/25 in.
Diameter (Metric) Inner 44.7 cm
Diameter (Metric) Outer 47 mm
Product Type Swagelok 1/16 in. Syringe
Max. Pressure 1500 psi
Volume (Metric) 200 mL
For Use With (Equipment) Syringe Pump
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.