Learn More
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Diameter (English) Inner | 1-19/25 in. |
| Diameter (English) Outer | 1-22/25 in. |
| Diameter (Metric) Inner | 44.7 cm |
| Diameter (Metric) Outer | 47 mm |
| Product Type | Swagelok 1/8 in. Syringe |
| Max. Pressure | 1500 psi |
| Volume (Metric) | 200 mL |
| For Use With (Equipment) | Syringe Pump |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.