Learn More
Bioss KD-Validated EOGT Rabbit Recom
SDP
Supplier:  Bioss BSM90084R
Encompass
EGF Domain Specific O-Linked N-Acetylglucosamine Transferase Primary Antibody Unconjugated Public Immunogen Range 1-150/527 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66837
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.