Learn More
Bioss EphA2 - Extracellular region M
SDP
Supplier:  Bioss BSM70481M
Encompass
EphA2 (Extracellular region) M088 Antibody Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications IP M088Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66538
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.