Learn More
Bioss KD-Validated UQCRC2 Rabbit Rec
SDP
Supplier:  Bioss BSM90489R
Encompass
Ubiquinol-Cytochrome C Reductase Core Protein 2 Primary Antibody Unconjugated Public Immunogen Range 1800-1865/1865 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.