Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDEL Receptor Pan Monoclonal Antibody (KR-10)
GREENER_CHOICE

Catalog No. PIMA190944
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190944 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190944 Supplier Invitrogen™ Supplier No. MA190944
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store as a concentrated solution. Centrifuge briefly prior to opening vial.

Retention of resident soluble proteins in the lumen of the endoplasmic reticulum (ER) is achieved in both yeast and animal cells by their continual retrieval from the cis-Golgi, or a pre-Golgi compartment. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually lys-asp-glu-leu (KDEL) in animal cells, and his-asp-glu-leu (HDEL) in S. cerevisiae. This process is mediated by a receptor that recognizes, and binds the tetrapeptide-containing protein, and returns it to the ER. In yeast, the sorting receptor encoded by a single gene, ERD2, which is a seven-transmembrane protein. Unlike yeast, several human homologs of the ERD2 gene, constituting the KDEL receptor gene family, have been described. The protein encoded by this gene was the first member of the family to be identified, and it encodes a protein structurally and functionally similar to the yeast ERD2 gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDEL Receptor Pan
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone KR-10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KDELR2
Gene Accession No. O43731, O76767, P24390, P33946, P33947, Q2KJ37, Q569A6, Q5U305, Q5ZKX9, Q7ZXS5, Q8R1L4, Q99JH8, Q9CQM2
Gene Alias (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; 1110007A14Rik; 8030486F04Rik; AI173274; AW215843; BEST:CK00230; CG5183; CG5183-PA; CG5183-PB; CG5183-PC; CK00230; Dmel\CG5183; Dmel_CG5183; dmErd2; ELP-1; ER lumem protein retaining receptor 2; ER lumen protein retaining receptor; ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 2; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor; ER lumen protein-retaining receptor 1; ER lumen protein-retaining receptor 2; ER lumen protein-retaining receptor 3; ERD2; Erd2.1; ERD2.2; ERD23; ERD2L3; ERD-2-like protein; ERD2-like protein 1; ergr15; HDEL; hypothetical protein; I79_001653; I79_020931; I79_023963; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 2; KDEL endoplasmic reticulum protein retention receptor 2 L homeolog; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor; KDEL receptor 1; KDEL receptor 2; KDEL receptor 3; KDEL receptor-like protein; KDELR; Kdelr1; Kdelr2; kdelr2.L; KDELR3; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; l(2)31Em; l(2)k00311; lethal (2) 31Em; PM23; Putative MAPK-activating protein PM23; RCJMB04_8l23; Unknown (protein for MGC:128955); XELAEV_18045328mg
Gene Symbols KdelR, KDELR1, KDELR2, kdelr2.L, KDELR3
Host Species Mouse
Immunogen Peptide corresponding to aa 192-212 of bovine KDEL receptor conjugated to KLH.
Purification Method Protein G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100338388, 100338871, 100349892, 100514523, 100520962, 100625446, 100750636, 100750912, 100765889, 101116624, 101121418, 101122047, 105785, 10945, 11014, 11015, 304290, 315131, 34427, 361577, 380330, 418026, 474509, 484406, 509687, 531676, 612839, 618184, 66913, 68137, 771927
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig, Chicken, Chicken, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.