Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM3A Monoclonal Antibody (OTI5A7), TrueMAB™, OriGene
SDP

Catalog No. 501684428
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4428 Supplier OriGene Supplier No. TA812138
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JMJD1A is a histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. It preferentially demethylates mono- and dimethylated H3 ′Lys-9′ residue, with a preference for dimethylated residue, while it has weak or no activity on trimethylated H3 ′Lys-9′. Demethylation of Lys residue generates formaldehyde and succinate. JMJD1A is involved in hormone-dependent transcriptional activation, by participating in recruitment to androgen-receptor target genes, resulting in H3 ′Lys-9′ demethylation and transcriptional activation. It′s also Involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 which are required for packaging and condensation of sperm chromatin. Involved in obesity resistance through regulation of metabolic genes such as PPARA and UCP1.

Specifications

Antigen KDM3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KDM3A
Gene Accession No. Q9Y4C1
Gene Alias JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
Gene Symbols KDM3A
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 850-1321 of human KDM3A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55818
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.