Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam KDM4/JMJD2 Activity Quantification Assay Kit, Fluorometric, 96 Tests

Catalog No. 502584897
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4897 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4897 Supplier Abcam Supplier No. AB11346296TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly measure KDM4/JMJD2 enzyme activity in 3 hours using this fluorometric assay kit.

  • Assay time: 3 hr
  • Assay type: Enzyme activity
  • Detection method: Fluorescent
  • Applications: Enzyme activity assay
  • Usage: Compatible with microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule KDM4A, JHDM3A, JMJD2A
No. of Reactions 96 Tests
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Format Kit
Quantity 96 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.