Learn More
KDM4C Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | KDM4C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene Alias | 2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; Gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; Jmjd2c; jumonji domain containing 2C; Jumonji domain-containing protein 2C; KDM4C; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; Lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against a peptide within human kDM4C. |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.