Learn More
Abcam KDM5/JARID Activity Quantification Assay Kit, Fluorometric, 48 Test
Description
- Assay Type: Enzyme activity
- Detection Method: Fluorescent
- Assay Time: 3 hr
- Applications: Enzyme activity assay
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | KDM5 (KDM5A/Jarid1A/RBBP2) |
| Run Time | 3 hr. |
| No. of Reactions | 48 Tests |
| Product Type | Western Blot Kit |
| Format | Kit |
| For Use With (Equipment) | Microplate Reader |
| Detection Method | Fluorescence |
| Quantity | 48 Tests |
| For Use With (Application) | Enzyme activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.