Learn More
Abcam KDM5/JARID Activity Quantification Assay Kit, Fluorometric, 96 Test
Description
- Assay Type: Enzyme activity
- Detection Method: Fluorescent
- Applications: Enzyme activity assay
- Assay Time: 3 hr
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| No. of Reactions | 96 Tests |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Run Time | 3 hr. |
| Target Molecule | KDM5A, Jarid1A, RBBP2 |
| Detection Method | Fluorescence |
| Quantity | 96 Tests |
| For Use With (Application) | Enzyme activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.