Learn More
Invitrogen™ KDM5D Polyclonal Antibody

Description
Antibody detects endogenous levels of total JAD1D.
Specifications
Specifications
| Antigen | KDM5D |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KDM5D |
| Gene Accession No. | Q9BY66 |
| Gene Alias | histocompatibility Y antigen; Histone demethylase JARID1D; HY; H-Y; HYA; JARID1D; Jumonji, AT rich interactive domain 1D (RBP2-like); jumonji/ARID domain-containing protein 1D; KDM5D; KIAA0234; lysine (K)-specific demethylase 5D; lysine demethylase 5D; lysine-specific demethylase 5D; Protein SmcY; selected mouse cDNA on Y, human homolog of; SMC homolog, Y chromosome; SMCY; Smcy homolog, Y-linked |
| Gene Symbols | KDM5D |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.