Learn More
Description
Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (93%), Xenopus laevis (82%), Dog (86%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | KDM6A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1612 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | KDM6A |
| Gene Accession No. | O15550 |
| Gene Alias | 4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
