Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR graduated laborator
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z23223810EA
Encompass
Reusable bottles without caps or pouring rings for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.Optional pouring rings prevent drips when pouring from these bottles.
Catalog No. 50-290-8680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.