Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR double walled jacke1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7423891EA
Encompass
The DURAN double walled wide mouth bottles GLSA80 incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN bottle. The DURAN double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.
Catalog No. 50-290-8789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.