Learn More
PALINTEST USA Palintest Kemio™ Disinfection ChloriteHard Case Kit
Supplier:  PALINTEST USA KEMH10DIS
Encompass_Preferred
USEPA approved for chlorite & chlorine dioxide and free & total chlorine measurement in drinking water.Powered by patented, single-use sensors which generate an electrical current by reactingwith the chemical in the water sample. Kemio measures this electrical signal to determinethe concentration of chemical or within the sample. With visual on-screen instructions and simple test method, potential user errors are minimized to ensure repeatable and reliable results. The integrated data log automatically captures 10,000 results to provide a traceable, auditable dataset. Kit contents: instrument, check standards, micro USB cable, international power adapter, stylus, hard carry case, 200 chlorine dioxide/chlorite sensors, 100 chlorine sensors, degassing equipment, CR-1 reagent, CR-2 reagent and glycine reagent.
Catalog No. NC1743037
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.