Learn More
PALINTEST USA Kemio™ Disinfection is a multiparameter testing platform. Tests for chlorine (free and total), chlorine dioxide, chlorite and peracetic acid (PAA) on one instrument with the use of different sensors.
Supplier: PALINTEST USA KEMS10DIS
Powered by patented, single-use sensors which generate an electrical current by reactingwith the chemical in the water sample. Kemio™ measures this electrical signal to determine the concentration of chemical or within the sample.Suitable for all sample types, Kemio™ delivers a consistent performance for samples which are colored, turbid or include floating particles.With visual on-screen instructions and simple test method, potential user errors are minimized to ensure repeatable and reliable results. The integrated data log automatically captures 10,000 results to provide a traceable, auditable dataset.Kit contents: instrument, check standards, micro USB, international power adaptor, stylus, inner protective carry case for instrument and outer soft carry case.*Sensors and reagents are ordered separately
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.