Learn More
Enzo Life Sciences Kemptide (1mg). CAS: 65189-71-1
SDP
Supplier:  Enzo Life Sciences BMLP1070001
Encompass_Preferred
Phosphate acceptor peptide. Substrate for Ser/Thr protein kinases. Km=4.7µM for cAMP-dependent protein kinase (PKA). The sequence defines the phosphorylation site of porcine liver pyruvate kinase. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1228
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.