Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ A Space Farm Kit
SCI_ED

Catalog No. S02080 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
4 to 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02080 4 to 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD—Small parts. Not for children under 3 yr.
Catalog No. S02080 Supplier Aqua Phoenix Scientific Supplier No. 1144
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOKING HAZARD—Small parts. Not for children under 3 yr.

Includes

Instructor manual, material safety data sheets, Petri dishes, Ziploc™ bags, plastic cups with lids, sunflower seeds, bean seeds, plastic toothpicks, space soil I, space soil II, pH paper, space salts, hydrochloric acid solution, sodium hydroxide solution

Requires

Two quart container and distilled water.

Illustrates to young students how we might grow our food in an environment other than earth.

  • Young scientists learn the needs of plants as they germinate seeds in nutrient solutions without soil
  • After experimenting with space salts and space soils, students may transfer seedlings to real soil from their own environment
  • Includes extensive information on hydroponic cultivation
  • Grades 4-6
  • Class size: 24 students

Specifications

Grade 4 to 6
For Use With (Application) Hydroponics
Includes Instructor manual, material safety data sheets, Petri dishes, Ziploc™ bags, plastic cups with lids, sunflower seeds, bean seeds, plastic toothpicks, space soil I, space soil II, pH paper, space salts, hydrochloric acid solution, sodium hydroxide solution
Class Size 24 Students
Description A Space Farm Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.