Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Accelerated Motion Kit
SCI_ED

Catalog No. S05897 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05897 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S05897 Supplier Aqua Phoenix Scientific Supplier No. 15104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

PVC Couplers, section of PVC pipe, 3-prong clamp extensions, right angle clamp, stopwatch, die cast car, metric tape, protractor, steel ball bearing, graph paper, instructor's manual, student data sheet

Requires

Impact goggles

Investigate accelerated motion using a totally reusable system that includes six foot tubular ramps, toy cars and ball bearings.

  • Simple apparatus with an exceptional support system that allows students to use timed trials to measure gravitational acceleration and the car's/ball's coefficient of friction
  • Adjustable system allows modification of track length and angle
  • Includes separate problem sets for general science and higher level Physics courses
  • Higher level curriculum includes kinetic and static coefficients of friction as well as bodies on inclined planes
  • Instructor's manual includes reproducible student instructions, guided calculations including graphing of data and calculation of error, assessment questions, sample data and answer keys
  • Appropriate for any level Physics course (including AP) as well as general science courses
  • Grades 9 to 12
  • Class size: 24 students

Specifications

Grade 8 to 12
For Use With (Application) Engages students in study of the basic laws of physics, kit investigates that phenomenon and relates it to a vector component of the acceleration due to gravity
Includes Instructor manual, student data sheet, pvc couplers, section of pvc pipe, 3-prong clamp extensions, right angle clamp, stopwatch, die cast car, metric tape, protractor, steel ball bearing, graph paper
Class Size 36 Students
Product Type Accelerated Motion Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.