Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Acids, Bases, and Salts Kit
SCI_ED

Catalog No. S193831 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students (6 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S193831 24 Students (6 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S193831 Supplier Aqua Phoenix Scientific Supplier No. 4458
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor Manual
  • Student Manuals
  • Student Quiz Sheets
  • MSDS
  • Retort Stands
  • Microplates (24 Well)
  • Microplates (96 Well)
  • Fine Tip Pipettes
  • Medicine Cups
  • Plastic Toothpicks
  • Universal Indicator Charts
  • Plastic Dropper Bottles
  • Sodium Hydroxide Solution
  • Acetic Acid Solution
  • Ammonium Hydroxide Solution
  • Acetic Acid Solution
  • Hydrochloric Acid Solution
  • Thymol Blue
  • Bromothymol Blue
  • Universal Indicator
  • Phenolphthalein Solution
  • Methyl Orange

Requires

Goggles and distilled water

Students engage in three experiments to investigate properties of acids, bases and salts.

Kit includes all required chemicals and supplies for performing experiments on a micro scale.

  • Five solution indicators are used to explore pH of acids and bases
  • Micro-titration procedure is used for reacting acids and bases to form salts and learn difference between strong and weak acids and bases
  • Students learn how titration curve shows endpoint or neutralization point for indicators
  • Students determine concentration of acetic acid in household vinegar by titrating the weak acid with a strong base and using a phenolphthalein indicator
  • Grades 9-12
  • Class size: 24 students

Specifications

Grade 8 to 12
For Use With (Application) For students to use five solution indicators to explore the ph of acids and bases
Includes Two bottles each of 1% and 0.1M Acetic Acid, Ammonium Hydroxide, Hydrochloric Acid, 0.1M and 0.01M Sodium Hydroxide, Vinegar, Bromothymol blue, Methyl Orange, Phenolphthalein and universal indicator, six large and six small microplates, 30 transfer pipets, 23 dropper bottles, six universal indicator color charts, six medicine cups, stirring sticks, retort stands, assessment manual, quiz sheets, detailed instruction manual
Class Size 24 Students (6 Groups)
Product Type Acids Bases and Salts Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.