Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Advanced Enzymology Kit
SCI_ED

Catalog No. s06758 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06758 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06758 Supplier Kemtec™ Supplier No. 8130
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Buffer pH 3 solution, buffer pH 4 solution, buffer pH 5 solution, buffer pH 6 solution, buffer pH 7 solution, buffer pH 8 solution, buffer pH 9 solution, buffer pH 10 solution, buffer substrate effects solution, buffer temperature effects solution, buffer inhibition solution, inhibitor solution, sucrose inhibitor, sucrose substrate effecis, sucrose temperature effects, sucrose pH substrate, 3,5 di-nitro salicylic solution (DNS), aniline solution, reply card for invertase enzyme and glucose solution, instructor manual, student data sheets, MSDS

Requires

Goggles, lab glassware including beakers, test tubes and graduated cylinders, hot plate, spectrophotometer

Explore enzyme kinetics with this comprehensive kit featuring buffers, substrates, inhibitors, and detailed experiments designed for advanced biology or chemistry students.

  • Complete mini-course covering enzyme kinetics and factors affecting enzyme activity
  • Includes pH buffers (pH 3 to pH 10), substrate and inhibitor solutions
  • Contains DNS reagent for measuring reducing sugars via color change
  • Experiments include effects of substrate concentration, competitive/non-competitive inhibitors, pH, and temperature on Invertase reaction rate
  • Comprehensive instructor manual and student data sheets included
  • Designed for classes up to 24 students (Grades 11 to 14)
  • Requires standard lab glassware, goggles, hot plate, and spectrophotometer (not included)
  • Includes reply card for ordering Invertase enzyme and glucose solutions separately (shipped only within continental U.S.)
  • MSDS sheets included for all chemicals provided

Specifications

Grade 10 to 14
For Use With (Application) Students can measure the amount of reducing sugars with the chemical dns which creates a distinctive color change
Includes Beakers, test tubes, graduated cylinders, hot plate, spectrophotometer, reply card, instructor manual, student data sheets, MSDS, Buffer pH 3, 4, 5, 6, 7, 8, 9, 10 solution, Buffer substrate effects solution, Buffer temperature effects solution, Buffer inhibition solution, inhibitor solution, Sucrose inhibitor, Sucrose substrate effecis, Sucrose temperature effects, Sucrose pH substrate, 3,5 Dinitrosalicylic solution, Aniline solution, invertase solution, standard Glucose solution
Requires Lab glassware, beakers, test tubes, graduated cylinders, hot plate, spectrophotometer
Class Size 24 Students
Product Type Enzymology Advanced Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.