Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AIDS Simutest Kit
SCI_ED

Catalog No. S06451 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
11 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06451 11 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06451 Supplier Aqua Phoenix Scientific Supplier No. 3403

missing translation for 'validMainframeMsg'

Includes

Serum samples A - H, plastic pipets, ELISA microtiter plates, antibody conjugate, chromagen, instructor manual, SDS

Requires

Goggles

Students run simulated ELISA tests for AIDS.

  • Students run eight patient samples in simulated ELISA to determine which patient(s) may have contracted the HIV virus
  • Includes simulated patient serums, conjugate, chromagen, and reusable treated ELISA microtiter plates
  • Totally safe for student use as it contains no blood serum or infectious diseases
  • Students experience the world of biotechnology and learn about HIV virus within this one hour lab investigation
  • For grades 11 to 14
  • Class size: 24 students

Choking hazard (1) Small parts. Not for children under 3 yrs.

Specifications

Grade 11 to 14
For Use With (Application) Hiv infection awareness
Includes Instructor manual, student manual, methods of assessment manual, student question sheets, msds, answer key, aids plates, pipets, distilled water, nitric acid solution 3m, nitric acid solution 1.5m, antibody conjugate, chromogen
Product Type AIDS Simutest Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.