Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Amazing Waves Kit
SCI_ED

Catalog No. S94959 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Amazing Waves Kit; Grades: 5 - 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S94959 Amazing Waves Kit; Grades: 5 - 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S94959 Supplier Aqua Phoenix Scientific Supplier No. 9116
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor Manual
  • Crayons
  • Marbles
  • Slinky
  • Sand, Ultra Fine
  • Sand, Fine
  • Sand, Coarse
  • Plastic Containers
  • Stopwatches
  • Wood Pieces
  • Rulers
  • Graduated Cylinders
  • Empty Bottles with caps
  • Food Coloring

Requires

Computers with internet connections for research projects and a document projector (optional)

Seven activities take students through examination and observation of waves, wave properties and influence of waves on coastline.

Special emphasis on Influence of Engineering, Technology and Science on Society and the Natural World; Human Impacts on Earth Systems; Ecosystem Dynamics, Functioning and Resilience; The Roles of Water in Earth's Surface Processes and Problem Solving

  • Three demonstrations (types of waves, wave creation and wave action)
  • Two hands on activities (regarding wave motion, sediments and beach erosion)
  • Research and presentation project, applying science and engineering to societal issues
  • Written assignment with art connections
  • Grades 5-9
  • Class size: 24 students

Specifications

Grade 5 to 9
For Use With (Application) Examination and observation of waves, wave properties and influence of waves on coastline
Includes Instructor Manual; Crayons; Marbles; Slinky; Sand, Ultra Fine; Sand, Fine; Sand, Coarse; Plastic Containers; Stopwatches; Wood Pieces; Rulers; Graduated Cylinders; Empty Bottles with caps; Food Coloring
Class Size 24 Students
Description Amazing Waves Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.