Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Anti A, Anti B Serum Set (5mL)
SCI_ED

Catalog No. s06447 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06447 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06447 Supplier Aqua Phoenix Scientific Supplier No. 3242
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Highly specific monoclonal antisera tested for HbsAg and HTLV-III (AIDS).

Standard laboratory practices should be used and all biological products should be treated as if capable of transmitting disease. Product is perishable and requires special shipping. Refrigerate for prolonged shelf life.

Kit Contains:

  • Two droppers
  • Anti-A serum, 5mL
  • Anti-B serum, 5mL
  • Approximately 36 tests
  • For grades 9 to 14

Specifications

Grade 8 to 12
Includes Two droppers, 5ml anti-a serum, 5ml anti-b serum
Product Type A and B Blood Typing Serum Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.