Learn More
Kemtec™ Anti-A, Anti-B, Serum Set, Lyophilized, 3-300L
SCI_ED
Catalog No. S06449 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 14
Catalog No. Grade
S06449 8 to 14
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06449 Supplier Aqua Phoenix Scientific Supplier No. 3300L
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.
Lyophilized sera allow prolonged, stable storage and shelf life.
Sera are tested for HbsAg and HTLV-III (AIDS), however standard laboratory practices should be used when handling and all biological products should be treated as if capable of transmitting disease. Refrigerate after reconstituting to extend shelf life.
Kit Contains:
  • Two droppers
  • Vials
  • Anti-A serum, lyophilized
  • Anti-B serum, lyophilized
  • Each set reconstitutes to 5mL Anti-A serum and 5 ml Anti-B serum with addition of sterile or distilled water
  • For grades 9 to 14
Specifications
Grade 8 to 14
Includes Two droppers, anti-a serum lyophilized, anti-b serum lyophilized
Product Type Anti A and Anti B Serum Set