Learn More
Kemtec™ Anti-Rh, Anti-D Serum, Lyophilized
SCI_ED
Catalog No. S06450 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 14
Catalog No. Grade
S06450 8 to 14
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06450 Supplier Aqua Phoenix Scientific Supplier No. 3302L
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.
Lyophilized sera allow prolonged, stable storage and shelf life.
Sera are tested for HbsAg and HTLV-III (AIDS), however standard laboratory practices should be used when handling and all biological products should be treated as if capable of transmitting disease. Refrigerate after reconstituting to extend shelf life.
Kit Contains:
  • Dropper
  • Vial
  • Anti-D (Rh) serum, lyophilized
  • Each vial reconstitutes to 5mL Anti-Rh serum with addition of sterile or distilled water
  • For grades 9 to 14
Specifications
Grade 8 to 14
Includes Dropper, anti-d serum lyophilized
Product Type Anti-Rh and Anti-D Serum Set