Learn More
Kemtec™ AP™ AP Chemistry Series Kits, 16 Total
SCI_ED
Catalog No. s07345 Shop All AquaPhoenix Scientific Products
Click to view available options
Product Type:
Basic AP Chemistry Series Kit
This item is not returnable. View return policy
Catalog No. S07345
Supplier: Kemtec™ AP™ 16130A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This bundle of 16 kits are designed to meet 2013 AP Next Generation Standards with inquiry-based labs and real-world chemistry applications.
  • Comprehensive Series: Includes 16 college-level AP chemistry labs, fully aligned with 2013 AP Next Generation National Standards
  • Kit Versions: Choose from Basic (no glassware) or Enhanced (includes glassware) versions
  • Engaging Curriculum: Hands-on investigations span stoichiometry, titrations, reaction kinetics, spectroscopy, thermochemistry, pKa, electrochemistry, and more
  • Technology Compatible: Works with TI-Nspire™ handhelds and software for optional data collection and simulations
  • Instructor Friendly: Designed for AP Chemistry classrooms with free digital activities available at fisheredu.com
Specifications
Certifications/Compliance Exceed 2013 AP next generation national standards
Grade 10 to 14
Includes Statistics, precision and accuracy, no Glassware, Spectrophotometric analysis of Copper, Beer's Law, synthesis and gravimetric analysis of Cobalt Oxalate Hydrate, Acid-Base titrations, an inquiry investigation, separating substances by absorption chromatography, stoichiometry, reactions with Copper compounds, stoichiometry, mole ratio of an unknown, determining molar mass by colligative properties, determining molar mass using the ideal gas equation, reaction rate and order, Determining molar enthalpy using Hess's law, Le Chatelier's principle featuring Ag+ reaction, identifying weak acids by pKa, preparing Buffer solutions and determining their properties, determining the equilibrium constant (and Temperature's Effect) with Beer's law, micro scale electrochemical cells voltaic and electrolytic
Product Type Basic AP Chemistry Series Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.