Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP Chemistry Kits: Determining the Equilibrium Constant (and Temperature's Effect) with Beer's Law
SCI_ED

Catalog No. S07343 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07343 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S07343 Supplier Aqua Phoenix Scientific Supplier No. 16117
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Iron (III) Nitrate, 0.25M, 400mL; Potassium Thiocyanate, 0.0025M, 275mL; Nitric Acid, 0.2N

Requires

Spectrophotometers, glassware and thermometers

Loaded with important concepts and techniques for the AP chemistry student; determine the equilibrium constant using Le Chatelier's principle and Beer's Law (using spectrophotometry) to determine equilibrium concentrations of reactants and products.

Addresses the inquiry question: What is the effect of temperature on the equilibrium constant?

  • Minimal prep time
  • Easily adapted to three shorter lab periods
  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 2 & 7
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 2 and 7
Grade 10 to 14
For Use With (Application) For the ap chemistry student, determine the equilibrium constant using le chatelier's principle and beer's law (using spectrophotometry) to determine equilibrium concentrations of reactants
Includes 0.25m, 400ml iron (iii) nitrate, 0.0025m, 275ml potassium thiocyanate, 0.2n nitric acid
Class Size 24 Students (12 Groups)
Product Type Determining the Equilibrium Constant (And Temperature'S Effect) with Beer'S Law Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.