Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Determining Molar Mass Using the Ideal Gas Equation
SCI_ED

Catalog No. s07337 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07337 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S07337 Supplier Aqua Phoenix Scientific Supplier No. 16109
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Isopropanol, ACS 99%; Methanol (Methyl Alcohol), ACS; Ethanol, 95% Denatured, 250mL

Requires

Hotplates, balances, ring stands, glassware and basic lab supplies

This unique hands-on method of exploring gases allows students to compare properties of real and ideal gases (based on KMT assumptions) as they determine the molar mass of a volatile solvent.

Students manipulate volume, temperature and pressure while using the ideal gas equation to compute moles of gas.

  • Ideal for Honors or AP Chemistry
  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 1, 2 & 5
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science Practices 1, 2 and 5
Grade 10 to 14
For Use With (Application) For students to compare properties of real and ideal gases (based on kmt assumptions) as they determine the molar mass of a volatile solvent, for honors or ap chemistry
Includes Isopropanol, acs 99%, methanol (methyl alcohol), acs, ethanol, 95% 250ml denatured
Class Size 24 Students (12 Groups)
Product Type Determining Molar Mass Using the Ideal Gas Equation Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.