Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Micro-scale Electrochemical Cells - Voltaic and Electrolytic
SCI_ED

Catalog No. S07344 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students (12 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07344 24 Students (12 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07344 Supplier Aqua Phoenix Scientific Supplier No. 16121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Copper Sulfate, 0.1M; Aluminum Sulfate, 0.1M, 60mL; Zinc Sulfate, 0.1M; Magnesium Sulfate 0.1M, 60mL; Ferric Sulfate 0.1M, 60mL; Potassium Chloride, 3.5M, 60mL; Potassium Iodide 0.5M; 12 Microplates, (24-Well); Six Copper Strips ( 2 in. x 1/4 in.); Six Aluminum Metal Strips; six Magnesium Strips (2 in. x 1/4 in.); Six Iron Strips (1/4 in. x 2.5 in.); Six Zinc Strips 2 in. x 1/4 in.); Six 9V Heavy Duty Batteries; Six I-Shape 9V Battery Snaps

Requires

DC voltmeter, glassware

Analyze oxidation-reduction (redox) reactions and become familiar with the activity series of metals and its importance in predicting the products of replacement reactions.

Students test both Voltaic and Electrolytic cells, and predict and measure cell voltages. Lessons include learning to separate oxidation-reduction (redox) reactions into half reactions, as well as identifying both anode and cathode activity.

 

  • Minimal prep time required
  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 3 & 7
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

 

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. 

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 3 and 7
Grade 10 to 14
For Use With (Application) To analyze oxidation-reduction reactions and become familiar with the activity series of metals and its importance in predicting the products of replacement reactions
Includes 0.1M Copper Sulfate, 0.1M, 60mL Aluminum Sulfate, 0.1M Zinc Sulfate, 0.1M, 60mL Magnesium Sulfate, 0.1M, 60mL Ferric Sulfate, 3.5M, 60mL Potassium Chloride, 0.5M Potassium Iodide, 12 Microplates, (24-Well), six Copper strips ( 2 in. x 1/4 in.), six Aluminum Metal strips, six Magnesium strips (2 in. x 0.25 in.), six Iron strips (0.25 in. x 2.5 in.), six Zinc strips 2 in. x 0.25 in.) six 9V Heavy duty batteries, six I-Shape 9V Battery snaps
Class Size 24 Students (12 Groups)
Product Type Micro-Scale Electrochemical Cells Voltaic and Electrolytic Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.