Learn More
Kemtec™ AP™ Chemistry Kits: Separating Substances by Absorption Chromatography
Shop All AquaPhoenix Scientific Products

Description
Includes
120mL ethanol, 120mL ethyl acetate, 3 silica gel, 1mL mixture of bromo brocreasol green and methyl red, 1mL zylene cyanl, 1mL mixture xylene cyanol brocreasol green and methyl red, 100 capillary tubes
Requires
24 well microtiter plates and basic laboratory supplies
Analyze various candies, foods, tea extracts and pen ink while comparing and contrasting paper chromatography and TLC results to reference materials. Teach students molecular polarity and intermolecular forces.
- Ideal for AP Chemistry, Chemistry I or II, or Honors Chemistry
- Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
- Kit designed to exceed 2013 AP Next Generation National Standards
- Meets AP Science Practices 1 & 5
- Targets Grades: 10-14
- Accommodates 24 students working in groups of two
Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire™ handhelds and software. Free activities can be found at www.fisheredu.com/TI.
Specifications
Specifications
| Grade | 10 to 14 |
| For Use With (Application) | Ideal for ap Chemistry, Chemistry i or ii, or Honors Chemistry |
| Includes | 120 mL ethanol, 120 mL ethyl acetate, 3 silica gel, 1 mL mixture of bromo brocreasol green and methyl red, 1 mL zylene cyanl, 1 mL mixture xylene cyanol brocreasol green and methyl red, 100 capillary tubes |
| Class Size | 24 Students |
| Product Type | Chromatography Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.