Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Statistics: Precision & Accuracy
SCI_ED

Catalog No. S07328 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students (12 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07328 24 Students (12 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S07328 Supplier Aqua Phoenix Scientific Supplier No. 16101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Analytical balance

Use a variety of basic volumetric measuring techniques to distinguish between precision and accuracy, while identifying an unknown liquid by its density.

Lessons include statistical analysis calculations of mean, standard deviation, and relative error. Students address the Inquiry Question: How is a physical constant used to verify the precision of a measuring tool? Ideal for AP Chemistry, Chemistry I or II, or Honors Chemistry.

  • Enhanced Kit includes glassware
  • Designed to exceed 2013 AP Next Generation National Standards.
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Meets AP Science Practice 2
  • Target Grades: 10 - 14
  • Accommodates 24 students working in groups of 2
Basic Kit Includes: Isopropanol, ACS 99%; Methanol (Methyl Alcohol), ACS; Ethyl Acetate, 250mL
Enhanced Kit Includes: Isopropanol, ACS 99%; Methanol (Methyl Alcohol), ACS; Ethyl Acetate, 250mL; 6 in. Glass Student Thermometer; Wire Test Tube Clamp; 50mL Heavyweight Erlenmeyer Flask; 50mL Beaker with White Markings; 10mL Hex based Cylinder; 10mL x 0.1 Disposable Poly Pipet; 3 in. Plastic Eyedropper; 50mm Poly Funnel

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire™ handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets AP Science Practice 2, exceed 2013 AP Next Generation National Standards
Grade 10 to 14
For Use With (Application) Basic volumetric measuring techniques to distinguish between precision and accuracy, while identifying an unknown liquid by its density
Includes Isopropanol, acs 99%, methanol (methyl alcohol), acs, ethyl acetate, 250ml
Class Size 24 Students (12 Groups)
Product Type Precision and Accuracy Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.