Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Stoichiometry: Reactions with Copper Compounds
SCI_ED

Catalog No. S07334 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07334 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07334 Supplier Aqua Phoenix Scientific Supplier No. 16106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Copper(II) Hydroxide, 15g; Copper Oxide, 15g; Sodium Acetate; Methanol (Methyl Alcohol), ACS; Sulfuric Acid, 6M; Aluminum Foil Sheet (12 x 10.75); 12 Plastic 5mL pipets

Requires

Fume hood, vacuum filtration (recommended), hotplates, balances and basic laboratory supplies

Perform four quantitatively different reactions utilizing copper compounds.

Identify reaction type, determine mole ratios experimentally, identify limiting reactants, and determine percent yield. The reaction series provides experience synthesizing ionic compounds, a coordination complex, and elemental copper.

  • Ideal for AP Chemistry or Honors Chemistry
  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 2 & 6
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire™ handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 2 and 6
Grade 10 to 14
For Use With (Application) For ap chemistry or honors chemistry
Includes 15g copper(ii) hydroxide, 15g copper oxide, sodium acetate, methanol (methyl alcohol), acs, 6m sulfuric acid, (12 x 10.75) aluminum foil sheet, 12 plastic 5ml pipets
Class Size 24 Students (12 Groups)
Product Type Stoichiometry Reactions with Copper Compounds Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.