Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Synthesis and Gravimetric Analysis of Cobalt Oxalate Hydrate
SCI_ED

Catalog No. s07331 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07331 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07331 Supplier Aqua Phoenix Scientific Supplier No. 16103

Includes

Oxalic Acid, 20g; Cobalt Chloride, 30g; Potassium Permanganate, 0.5N; Ammonium Hydroxide, Tech; Sulfuric Acid,12.0N (6M)

Requires

Analytical balance, burets, Bunsen burners, standard laboratory glassware

Students will enjoy this lab as they experience multiple techniques required for AP chemistry; synthesizing cobalt oxalate hydrate by precipitation, gravimetric analysis by dehydration, and oxidation-reduction (redox) titration on a unique compound.

  • Experiments can be easily adjusted to fit your lab period
  • Minimal prep time
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Meets AP Science Practices 2 & 7
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of 2

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire™ handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards
Grade 10 to 14
For Use With (Application) For students to enjoy this lab as they experience multiple techniques required for ap chemistry, synthesizing cobalt oxalate hydrate by precipitation, gravimetric analysis by dehydration, and oxidation-reduction titration on a unique compound
Includes 20g oxalic acid, 30g cobalt chloride, 0.5n potassium permanganate, ammonium hydroxide, tech, 12.0n (6m) sulfuric acid
Class Size 24 Students (12 Groups)
Product Type Synthesis and Gravimetric Analysis of Cobalt Oxalate Hydrate Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.